MilliporeSigma
  • Safety aspects of longitudinal administration of IGF-I/IGFBP-3 complex in neonatal mice.
Safety aspects of longitudinal administration of IGF-I/IGFBP-3 complex in neonatal mice.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society (2011-06-24)
Gunnel Hellgren, Wei Han, Xiaoyang Wang, Chatarina Löfqvist, Henrik Hagberg, Carina Mallard, Ann Hellström
ABSTRACT
Very preterm birth is associated with a high risk of morbidity. Infants born very preterm have low serum levels of insulin-like growth factor I (IGF-I), that further decrease after birth. IGF-I is essential for brain development and low serum levels have been associated with retinopathy of prematurity. The present study aimed to investigate the effects of prolonged administration of a low dose of rhIGF-I/rhIGFBP-3 on glucose levels and total body weight, as well as liver, spleen and brain weights, and gray and subcortical white matter in newborn mice. The study was performed as three different trials. In all experiments C57BL/6N mice were injected with a rhIGF-I/rhIGFBP-3 complex or saline. In the first experimental trial, blood glucose levels were assessed 30 min, 1 h, 1.5 h, 3 h, 6 h, 24 h and 48 h after the rhIGF-I/rhIGFBP-3 or saline injection on postnatal day (PND) 6. In the second trial, mice were injected daily from PND 3 to 11 and sacrificed on PND 12 for analysis of IGF-I serum levels. In the third trial, body and organ weights and effects on gray and white matter were assessed on PND 18 after PND 3-11 treatments as above. Effects on gray and white matter were measured using immunoreactivity for microtubule-associated protein-2 (MAP-2), myelin basic protein (MBP), 2',3'-cyclic nucleotide 3' phosphodiesterase (CNPase), neurofilament and oligodendrocyte lineage transcription factor 2 (Olig2). Blood glucose levels were unchanged in the rhIGF-I/rhIGFBP-3-treated group compared to baseline. In the control group glucose levels increased 30 min after the second saline injection; levels were not elevated at the subsequent time point. Three hours after the rhIGF-I/rhIGFBP-3 or saline, glucose levels were lower in rhIGF-I/rhIGFBP-3-treated animals than in saline treated (p=0.026). At PND 18, total body weight was higher in rhIGF-I/rhIGFBP-3-treated mice compared with controls (p<0.05), but there were no differences between groups in brain, liver or spleen weights. No differences in gray matter area were found between groups. Analyses of white matter markers showed an increased number of Olig2-positive cells in rhIGF-I/rhIGFBP-3-treated mice compared with controls (p<0.001). There were no differences between groups in terms of MBP, CNPase or neurofilament immunoreactivity. Prolonged administration of rhIGF-I/rhIGFBP-3 did not have a negative impact on blood glucose levels and was beneficial for total body growth.