MilliporeSigma
  • Neuroprotection associated with running: is it a result of increased endogenous neurotrophic factors?
Neuroprotection associated with running: is it a result of increased endogenous neurotrophic factors?
Neuroscience (2003-04-18)
E T Ang, P T H Wong, S Moochhala, Y K Ng
ABSTRACT
The possible neuroprotective effect of physical exercise was investigated in rats after middle cerebral artery occlusion (MCAO), a focal stroke model. It was found that physical exercise in the form of a 12-week treadmill running programme reduced the volume of infarction caused by MCAO. At the molecular level, reverse transcription polymerase chain reaction revealed that the runner had increased gene expression for nerve growth factor (NGF) over the nonrunner with or without MCAO. Expression of the NGF receptors, p75, was increased only in the absence of MCAO. In addition, runners showed a significantly higher number of cholinergic neurons, which constitutively expressed p75, in the horizontal diagonal band of Broca. The present findings suggest that neuroprotection after physical exercise may be a result of an increase in an endogenous neurotrophic factor nerve growth factor and the proliferation of its receptive cholinergic neurons.