MilliporeSigma
  • Delayed NGF infusion fails to reverse axotomy-induced degeneration of basal forebrain cholinergic neurons in adult p75(LNTR)-deficient mice.
Delayed NGF infusion fails to reverse axotomy-induced degeneration of basal forebrain cholinergic neurons in adult p75(LNTR)-deficient mice.
Neuroscience (2002-04-06)
C E E M van der Zee, T Hagg
ABSTRACT
The p75 low-affinity neurotrophin receptor (p75(LNTR)) appears to have various functions that include enhancing nerve growth factor (NGF)-mediated survival by increasing TrkA (high-affinity NGF receptor) efficiency, and mediating apoptosis by acting as a ligand-regulated pro-apoptotic receptor. Here, we investigated the role of p75(LNTR) for adult cholinergic basal forebrain neurons by comparing neuronal responses to injury in control and p75(LNTR)-deficient mice. In both types of mice, approximately 70% of the cholinergic neurons in the ipsilateral medial septum had lost their markers choline acetyltransferase and tyrosine kinase A by 28 days following unilateral transection of the dorsal septohippocampal pathway (fimbria fornix). A 7-day delayed infusion of NGF that started 28 days after the injury resulted in reversal of choline acetyltransferase expression and cell atrophy in control, but not in p75(LNTR)-deficient, mice. This lack of response to delayed NGF treatment in p75(LNTR)-deficient mice was most likely not due to cell death, as all of the septohippocampal neurons, labeled with Fluorogold before the lesion, were present at 28 days post-lesion, similar to control mice. p75(LNTR)-deficient cholinergic neurons can respond to NGF as they were protected by NGF infusions that started immediately after the injury. These observations, the fact that lesioned p75(LNTR)-deficient neurons atrophy faster, and that non-lesioned neurons hypertrophy in response to NGF in control but not in p75(LNTR)-deficient mice, suggest that p75(LNTR) is needed for tyrosine kinase A and NGF signaling efficiency.In conclusion, during adulthood p75(LNTR) appears to play a beneficial role in the response of cholinergic neurons to injury, consistent with the proposed role of p75(LNTR) in the enhancement of TrkA signaling and the transport of neurotrophins by these neurons.