MilliporeSigma
  • Redox state is a central modulator of the balance between self-renewal and differentiation in a dividing glial precursor cell.
Redox state is a central modulator of the balance between self-renewal and differentiation in a dividing glial precursor cell.
Proceedings of the National Academy of Sciences of the United States of America (2000-08-16)
J Smith, E Ladi, M Mayer-Proschel, M Noble
ABSTRACT
We have discovered that intracellular redox state appears to be a necessary and sufficient modulator of the balance between self-renewal and differentiation in dividing oligodendrocyte-type-2 astrocyte progenitor cells. The intracellular redox state of freshly isolated progenitors allows prospective isolation of cells with different self-renewal characteristics. Redox state is itself modulated by cell-extrinsic signaling molecules that alter the balance between self-renewal and differentiation: growth factors that promote self-renewal cause progenitors to become more reduced, while signaling molecules that promote differentiation cause progenitors to become more oxidized. Moreover, pharmacological antagonists of the redox effects of these cell-extrinsic signaling molecules antagonize their effects on self-renewal and differentiation, indicating that cell-extrinsic signaling molecules that modulate this balance converge on redox modulation as a critical component of their effector mechanism.