MilliporeSigma
Zinc-rich synaptic boutons in human temporal cortex biopsies.
Neuroscience (2000-06-28)
N Franco-Pons, C Casanovas-Aguilar, S Arroyo, J Rumià, J Pérez-Clausell, G Danscher
ABSTRACT
The distribution of zinc-rich synaptic boutons in biopsies of the temporal cortex from epileptic patients who had undergone surgery is described. Unfixed cryostat sections were exposed to H(2)S vapour to precipitate endogenous zinc, which was subsequently shown by silver enhancement. In the temporal cortex, the stain for zinc was arranged in bands: stain was heavy in layers II and VI, moderate-to-heavy in layers I, III and V, and low in layer IV. The white matter was virtually devoid of staining. At the electron microscope level, labelling was found in synaptic boutons that made asymmetric synaptic contacts. Immunohistochemical staining for glutamate receptor subunits GluR2/3 was observed in cell bodies in layers II, III, V and VI, coincident with the layers that showed heavy staining for zinc. Immunostaining for glutamate receptor subunit GluR1 was prominent in non-pyramidal neurons in deep cortical layers. These results support findings in other mammals and indicate that the human neocortex may contain an extensive system of zinc-rich cortico-cortical connections. This system may be altered in pathological conditions.