MilliporeSigma
  • Halophilic lipase does forms catalytically active aggregates: Evidence from Marinobacter sp. EMB5 lipase (LipEMB5).
Halophilic lipase does forms catalytically active aggregates: Evidence from Marinobacter sp. EMB5 lipase (LipEMB5).
International journal of biological macromolecules (2018-07-22)
R Hemamalini, Sunil Kumar Khare
ABSTRACT
Lipases are prone to aggregation due to the presence of hydrophobic surface and corresponding hydrophobic interactions. However, like most lipases, halophilic lipases do not tend to aggregate owing to the presence of charged surface. Conversely, Marinobacter sp. EMB5 produces a unique lipase (LipEMB5) which tends to form functionally active aggregate, despite a presence of 2% (w/v) NaCl in the environment. Gel filtration using Sephacryl S-200 column resulted in elution of lipase in the void volume despite a high salt level, suggesting aggregation behavior. The aggregated form of LipEMB5 (172.35kDa) was shown to have specific activity of 16.3IUmg-1 protein. The disaggregated form (<6kDa) with increased elution volume was also obtained by inclusion of 70% 2-propanol in the elution buffer. It was catalytically less active (0.086IUmg-1 protein specific activity) in comparison to aggregated form. Aggregated and 2-propanol induced disaggregated forms were characterized in terms of DLS, biochemical, morphological, and structural properties. Broadly, the aggregated form showed 24 times more activity with 1000-fold less Km compared to its counterpart. The enzymatic properties were almost observed to be same for both the species. However, better structural integrity in the disaggregated form was observed on the basis of FTIR and CD studies. Overall, the study comes out with a unique halophilic lipase from Marinobacter sp. EMB5 which have a tendency to form active aggregates in high salt laden environment.
Product Description
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 392 mPa.s (25 °C)