MilliporeSigma
  • Effect of integrin α5β1 inhibition on SDF-l/CXCR4-mediated choroidal neovascularization.
Effect of integrin α5β1 inhibition on SDF-l/CXCR4-mediated choroidal neovascularization.
International journal of ophthalmology (2018-06-05)
Yang Lyu, Wen-Qin Xu, Li-Juan Sun, Xiao-Yan Pan, Jian Zhang, Yu-Sheng Wang
ABSTRACT
To investigate the roles of integrins in choroidal neovascularization (CNV) and their associations with the stromal cell-derived factor-1 (SDF-1)/CXCR4 axis. CNV lesions were induced in mice using laser photocoagulation. After CNV induction, all animals were randomly assigned to: control, SDF-1, SDF-1+age-related macular degeneration (AMD) 3100 (CXCR4 inhibitor), and SDF-1+ATN161 (integrin α5β1 inhibitor) groups; their effects on CNV progression were observed using hematoxylin eosin (HE) staining, fundus fluorescein angiography (FFA) grading and optical coherence tomography (OCT), and their effects on CXCR4/integrin α5 expression were evaluated using Western blot and double immunofluorescence staining. Hypoxia-exposed endothelial cells (ECs) were used to simulate CNV in vitro, they were treated with SDF-1, combined with CXCR4 siRNA/AMD3100 or ATN161, and expression of integrin α5, cell migration and tube formation were analyzed. Integrin subunit α5 increased at 3rd and 7th day and decreased at 14th day in CNV mice, with no significant change of β1-integrin. CXCR4 expression in CNV mice had persistent increase within 14d after induction. SDF-1 treatment significantly promoted the CNV progression during 3-14d. The mean CNV length in AMD3100 and ATN161 group at day 7 was 270.13 and 264.23 µm in HE images, significantly lower than the mean length in SDF-1 (345.70 µm) group. AMD3100 and ATN161 also significantly reduced thickness and leakage of CNV induced by SDF-1. Mean integrin α5 positive area in SDF-1 group reached 2.31×104 µm2, significantly higher than control (1.25×104 µm2), which decreased to 1.78×104 µm2 after AMD3100 treatment. About 61.36% of ECs in CNV lesions expressed α5 in SDF-1 group, which significantly decreased to 43.12% after AMD3100 treatment. In vitro, integrin α5 peaked by 6 folds after 6h of hypoxia exposure and CXCR4 gradually increased by up to 2.3 folds after 24h of hypoxia. Approximately 25.12% of ECs expressed integrin α5 after SDF-1 stimulation, which decreased to 7.2%-9.5% after si-CXCR4 or AMD3100 treatment. ATN161 exerted an inhibitory effect comparable to that of si-CXCR4 on EC migration and tube formation in the presence of SDF-1. SDF-1/CXCR4 signaling induces integrin α5β1 expression in ECs to promote CNV.