MilliporeSigma
  • Single-cell analysis of normal and FOXP3-mutant human T cells: FOXP3 expression without regulatory T cell development.
Single-cell analysis of normal and FOXP3-mutant human T cells: FOXP3 expression without regulatory T cell development.
Proceedings of the National Academy of Sciences of the United States of America (2006-04-18)
Marc A Gavin, Troy R Torgerson, Evan Houston, Paul DeRoos, William Y Ho, Asbjørg Stray-Pedersen, Elizabeth L Ocheltree, Philip D Greenberg, Hans D Ochs, Alexander Y Rudensky
ABSTRACT
Forkhead winged-helix transcription factor Foxp3 serves as the dedicated mediator of the genetic program governing CD25+CD4+ regulatory T cell (T(R)) development and function in mice. In humans, its role in mediating T(R) development has been controversial. Furthermore, the fate of T(R) precursors in FOXP3 deficiency has yet to be described. Making use of flow cytometric detection of human FOXP3, we have addressed the relationship between FOXP3 expression and human T(R) development. Unlike murine Foxp3- T cells, a small subset of human CD4+ and CD8+ T cells transiently up-regulated FOXP3 upon in vitro stimulation. Induced FOXP3, however, did not alter cell-surface phenotype or suppress T helper 1 cytokine expression. Furthermore, only ex vivo FOXP3+ T(R) cells persisted after prolonged culture, suggesting that induced FOXP3 did not activate a T(r) developmental program in a significant number of cells. FOXP3 flow cytometry was also used to further characterize several patients exhibiting symptoms of immune dysregulation, polyendocrinopathy, enteropathy, X-linked syndrome (IPEX) with or without FOXP3 mutations. Most patients lacked FOXP3-expressing cells, further solidifying the association between FOXP3 deficiency and immune dysregulation, polyendocrinopathy, enteropathy, X-linked syndrome. Interestingly, one patient bearing a FOXP3 mutation enabling expression of stable FOXP3(mut) protein exhibited FOXP3(mut)-expressing cells among a subset of highly activated CD4+ T cells. This observation raises the possibility that the severe autoimmunity in FOXP3 deficiency can be attributed, in part, to aggressive T helper cells that have developed from T(R) precursors.