Gluconolactone
Synonym(s):
D-(+)-Gluconic acid δ-lactone, δ-Gluconolactone, 1,2,3,4,5-Pentahydroxycaproic acid δ-lactone, D-(+)-Dextronic acid δ-lactone
Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Application
Package Size
Special Grade
Showing 1-2 of 2