Skip to Content
MilliporeSigma
β-Nicotinamide adenine dinucleotide hydrate ≥98%, BioUltra, from yeast

β-Nicotinamide adenine dinucleotide hydrate

β-Nicotinamide adenine dinucleotide hydrate ≥98%, BioUltra, from yeast
Synonym(s):
β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide, β-Nicotinamide adenine dinucleotide hydrate, Nicotinamide adenine dinucleotide free acid, NAD+, Free Acid
Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)

Products

Product Category
Brand
Sustainability
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
pH Value
Physical Form
Purity
Research Area
Quality Segment
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing