MilliporeSigma
  • Identification of novel serum autoantibodies against EID3 in non-functional pancreatic neuroendocrine tumors.
Identification of novel serum autoantibodies against EID3 in non-functional pancreatic neuroendocrine tumors.
Oncotarget (2018-01-02)
Koji Hontani, Takahiro Tsuchikawa, Takaki Hiwasa, Toru Nakamura, Takashi Ueno, Toshihiro Kushibiki, Mizuna Takahashi, Kazuho Inoko, Hironobu Takano, Satoshi Takeuchi, Hirotoshi Dosaka-Akita, Masaki Kuwatani, Naoya Sakamoto, Yutaka Hatanaka, Tomoko Mitsuhashi, Hideaki Shimada, Toshiaki Shichinohe, Satoshi Hirano
ABSTRACT
Pancreatic neuroendocrine tumors (pNETs) are relatively rare heterogenous tumors, comprising only 1-2% of all pancreatic neoplasms. The majority of pNETs are non-functional tumors (NF-pNETs) that do not produce hormones, and as such, do not cause any hormone-related symptoms. As a result, these tumors are often diagnosed at an advanced stage because patients do not present with specific symptoms. Although tumor markers are used to help diagnosis and predict some types of cancers, chromogranin A, a widely used tumor marker of pNETs, has significant limitations. To identify novel NF-pNET-associated antigens, we performed serological identification of antigens by recombinant cDNA expression cloning (SEREX) and identified five tumor antigens (phosphatase and tensin homolog, EP300-interacting inhibitor of differentiation 3 [EID3], EH domain-containing protein 1, galactoside-binding soluble 9, and BRCA1-associated protein). Further analysis using the AlphaLISA