MilliporeSigma
  • Computational detection and quantification of human and mouse neutrophil extracellular traps in flow cytometry and confocal microscopy.
Computational detection and quantification of human and mouse neutrophil extracellular traps in flow cytometry and confocal microscopy.
Scientific reports (2017-12-21)
Brandon G Ginley, Tiffany Emmons, Brendon Lutnick, Constantin F Urban, Brahm H Segal, Pinaki Sarder
ABSTRACT
Neutrophil extracellular traps (NETs) are extracellular defense mechanisms used by neutrophils, where chromatin is expelled together with histones and granular/cytoplasmic proteins. They have become an immunology hotspot, implicated in infections, but also in a diverse array of diseases such as systemic lupus erythematosus, diabetes, and cancer. However, the precise assessment of in vivo relevance in different disease settings has been hampered by limited tools to quantify occurrence of extracellular traps in experimental models and human samples. To expedite progress towards improved quantitative tools, we have developed computational pipelines to identify extracellular traps from an in vitro human samples visualized using the ImageStream