MilliporeSigma
  • Red Blood Cell Passage of Small Capillaries Is Associated with Transient Ca
Red Blood Cell Passage of Small Capillaries Is Associated with Transient Ca
Frontiers in physiology (2017-12-21)
Jens G Danielczok, Emmanuel Terriac, Laura Hertz, Polina Petkova-Kirova, Franziska Lautenschläger, Matthias W Laschke, Lars Kaestner
ABSTRACT
When red blood cells (RBCs) pass constrictions or small capillaries they need to pass apertures falling well below their own cross section size. We used different means of mechanical stimulations (hypoosmotic swelling, local mechanical stimulation, passing through microfluidic constrictions) to observe cellular responses of human RBCs in terms of intracellular Ca
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid