MilliporeSigma
  • Annexin A2-modulated proliferation of pulmonary arterial smooth muscle cells depends on caveolae and caveolin-1 in hepatopulmonary syndrome.
Annexin A2-modulated proliferation of pulmonary arterial smooth muscle cells depends on caveolae and caveolin-1 in hepatopulmonary syndrome.
Experimental cell research (2017-07-22)
Lin Liao, Binwu Zheng, Bin Yi, Chang Liu, Lin Chen, Ziyang Zeng, Jing Gao
ABSTRACT
We have established that annexin A2 (ANXA2) is an important factor in the experimental hepatopulmonary syndrome (HPS) serum-induced proliferation of pulmonary arterial smooth muscle cells (PASMCs). However, the detailed mechanism remains unclear. ANXA2 translocated to the caveolin-enriched microdomains (caveolae) in PASMCs upon HPS serum stimulation. The disruption of caveolae by Methyl-β-cyclodextrin (MβCD) alleviated the caveolae recruitment of ANXA2 and the ANXA2-mediated activation of ERK1/2 and NF-κB, so that ANXA2-modulated PASMC proliferation was suppressed. The over-expression of Cav-1 resulted in the relocation of ANXA2 from caveolae and negatively regulated ERK1/2 and NF-κB activation, which inhibited the ANXA2-modulated PASMC proliferative behavior. These data indicate that caveolae function as a signaling platform for ANXA2-induced proliferative behavior and Cav-1 participates upstream of ANXA2 in the activation of ERK1/2 and NF-κB.