MilliporeSigma
Identification of residues that control Li
Biochimica et biophysica acta (2017-01-29)
Soumitra Roy, Kuntal Dey, Michal Hershfinkel, Ehud Ohana, Israel Sekler
ABSTRACT
The Na We combined molecular modeling to map transport site of NCLX with euryarchaeal H Mutation of Asn149, Pro152, Asp153, Gly176, Asn467, Ser468, Gly494 and Asn498 partially or strongly abolished mitochondrial Ca We identified distinct Na The results of this study provide functional insights into the unique Li