MilliporeSigma
Effect of Gold Nanoparticle Distribution in TiO
Nanoscale research letters (2017-08-31)
Shuichi Mayumi, Yutaka Ikeguchi, Daisuke Nakane, Yasuaki Ishikawa, Yukiharu Uraoka, Mamoru Ikeguchi
ABSTRACT
Photoanodes comprising Au nanoparticles (GNPs) and thin TiO
Sigma-Aldrich
4-tert-Butylpyridine, 98%