MilliporeSigma
  • 1,2,3,4,6-Penta-O-galloyl-ß-D-glucose, a bioactive compound in Elaeocarpus sylvestris extract, inhibits varicella-zoster virus replication.
1,2,3,4,6-Penta-O-galloyl-ß-D-glucose, a bioactive compound in Elaeocarpus sylvestris extract, inhibits varicella-zoster virus replication.
Antiviral research (2017-07-03)
Sohee Bae, Sun Yeou Kim, Moon Ho Do, Chan Hee Lee, Yoon-Jae Song
ABSTRACT
The aim of this study was to establish the effect of a 70% ethanol extract of Elaeocarpus sylvestris (ESE) on varicella-zoster virus (VZV) replication and identify the specific bioactive component(s) underlying its activity. ESE induced a significant reduction in replication of the clinical strain of VZV. Activity-guided fractionation indicated that the ethyl acetate (EtOAc) fraction of ESE contains the active compound(s) inhibiting VZV replication. High-Performance Liquid Chromatography coupled to Electrospray Ionization Quadrupole Time-of-Flight Mass Spectrometry (HPLC-Q-TOF-MS/MS) analysis of the EtOAc fraction of ESE facilitated the identification of 13 chemical components. Among these, 1,2,3,4,6-penta-O-galloyl-ß-D-glucose (PGG) markedly suppressed VZV-induced c-Jun N-terminal kinase (JNK) activation, expression of viral immediate-early 62 (IE62) protein and VZV replication. Our results collectively support the utility of PGG as a potential candidate anti-viral drug to treat VZV-associated diseases.
Sigma-Aldrich
Penta-O-galloyl-β-D-glucose hydrate, ≥96% (HPLC)