MilliporeSigma
  • Feather corticosterone during non-breeding correlates with multiple measures of physiology during subsequent breeding in a migratory seabird.
Feather corticosterone during non-breeding correlates with multiple measures of physiology during subsequent breeding in a migratory seabird.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2017-03-01)
Graham D Fairhurst, Louise Champoux, Keith A Hobson, Jean-François Rail, Jonathan Verreault, Magella Guillemette, William A Montevecchi, Pauline Brousseau, Catherine Soos
ABSTRACT
Carry-over effects in migratory birds are likely mediated by physiological processes that are activated in response to environmental variation. Such processes affect body condition and/or reproductive success, and can include corticosterone (CORT) because this hormone responds to environmental stressors and influences energy balance. Few studies have considered how CORT levels during non-breeding relate to a broader physiological profile during subsequent breeding, and fewer still have considered measures other than body condition. To explore CORT's potential role in carry-over effects, we investigated the relationship between CORT and foraging ecology of northern gannets (Morus bassanus) during the non-breeding period, and tested for associations between these factors and variation in a suite of physiological and biochemical metrics during subsequent breeding. Northern gannets are the largest seabird top predator in the North Atlantic and were among the hardest hit by the Deepwater Horizon oil blowout in the Gulf of Mexico in 2010. We used light-level geolocators to confirm winter origins of individuals in our study. No interrelationships were found among levels of CORT from feathers grown during non-breeding (CORT