MilliporeSigma
  • Monocytic thrombomodulin promotes cell adhesion through interacting with its ligand, Lewis
Monocytic thrombomodulin promotes cell adhesion through interacting with its ligand, Lewis
Immunology and cell biology (2016-11-04)
Wei-Ling Lin, Chia-Chi Chen, Guey-Yueh Shi, Chih-Yuan Ma, Chuan-Fa Chang, Hua-Lin Wu
ABSTRACT
The leukocyte adhesion cascade involves multiple events that efficiently localize circulating leukocytes into the injured sites to mediate inflammatory responses. From rolling to firm adhesion, the interactions between adhesion molecules have pivotal roles in increasing the avidity of leukocytes to endothelial cells. Thrombomodulin (TM), an essential anticoagulant protein in the vasculature, is also expressed on leukocytes. We previously demonstrated that Lewis