MilliporeSigma
  • Effect of epigallocatechin gallate on growth performance and antioxidant capacity in heat-stressed broilers.
Effect of epigallocatechin gallate on growth performance and antioxidant capacity in heat-stressed broilers.
Archives of animal nutrition (2017-07-26)
Bo Xue, Jiao Song, Longzhou Liu, Jingxian Luo, Guangming Tian, Ye Yang
ABSTRACT
This study investigated the effects of epigallocatechin gallate (EGCG) on the growth performance and antioxidant capacity of 35-d-old broilers exposed to heat stress. Broilers, 14 d of age, were divided into four groups with six replicates per group (eight chickens/replicate). Thermoneutral group (Group TN) was fed the basal diet and maintained at 28°C for 24 h/d. The heat-stressed groups were housed at 35°C for 12 h/d and 28°C for 12 h/d and fed the basal diet supplemented with EGCG at 0, 300 and 600 mg/kg diet (Groups HS0, HS 300 and HS600, respectively). Compared with Group TN, heat-stressed groups showed significantly reduced gain, feed intake and serum total protein and glucose levels; inhibited serum alkaline phosphatase activities; and increased serum levels of uric acid, cholesterol and triglycerides and the activity of serum creatine kinase, lactate dehydrogenase and aspartate aminotransferase (p < 0.05). Compared with Group HS0, Group HS600 exhibited an increased gain and feed intake; and normalised blood parameters and enzyme activities. Compared with Group TN, the expression of antioxidant-related liver proteins was decreased in Group HS0 and increased in Groups HS300 and HS600 (p < 0.05). The results suggest that EGCG can improve the growth performance and alleviate the oxidant damage by modulating the antioxidant properties of broilers.