MilliporeSigma
  • Triterpenoids-Enriched Extract from the Aerial Parts of Salvia miltiorrhiza Regulates Macrophage Polarization and Ameliorates Insulin Resistance in High-Fat Fed Mice.
Triterpenoids-Enriched Extract from the Aerial Parts of Salvia miltiorrhiza Regulates Macrophage Polarization and Ameliorates Insulin Resistance in High-Fat Fed Mice.
Phytotherapy research : PTR (2016-10-08)
Jing Leng, Mei-Hong Chen, Zhi-Hui Zhou, Ya-Wen Lu, Xiao-Dong Wen, Jie Yang
ABSTRACT
Adipose tissue inflammation and macrophage polarization are tightly associated with the development of obesity-associated insulin resistance. Our previous studies have demonstrated the triterpenoids-enriched extract from the aerial parts of Salvia miltiorrhiza (TTE) could significantly improve atherosclerosis in LDLR