MilliporeSigma
  • Array-CGH revealed one of the smallest 16q21q22.1 microdeletions in a female patient with psychomotor retardation.
Array-CGH revealed one of the smallest 16q21q22.1 microdeletions in a female patient with psychomotor retardation.
European journal of paediatric neurology : EJPN : official journal of the European Paediatric Neurology Society (2013-01-29)
Eirini Tsoutsou, Maria Tzetis, Krinio Giannikou, Areti Syrmou, Vasilis Oikonomakis, Konstantina Kosma, Anastasia Kanioura, Emmanuel Kanavakis, Helen Fryssira
ABSTRACT
A 28-month-old girl with dysmorphic craniofacial features, microcephaly, hypotonia, psychomotor retardation, failure to thrive and gastrointestinal problems was referred for clinical evaluation. Array-CGH analysis revealed one of the smallest de novo microdeletions on chromosome 16q21q22.1, 2.03 Mb in size. Advanced molecular analysis contributes to more precise genotype-phenotype correlation and accurate definition of the breakpoints in the deleted/duplicated regions.