MilliporeSigma
  • CD300f:IL-5 cross-talk inhibits adipose tissue eosinophil homing and subsequent IL-4 production.
CD300f:IL-5 cross-talk inhibits adipose tissue eosinophil homing and subsequent IL-4 production.
Scientific reports (2017-07-21)
Perri Rozenberg, Hadar Reichman, Israel Zab-Bar, Michal Itan, Metsada Pasmanik-Chor, Carine Bouffi, Udi Qimron, Ido Bachelet, Patricia C Fulkerson, Marc E Rothenberg, Ariel Munitz
ABSTRACT
Eosinophils and their associated cytokines IL-4 and IL-5 are emerging as central orchestrators of the immune-metabolic axis. Herein, we demonstrate that cross-talk between the Ig-superfamily receptor CD300f and IL-5 is a key checkpoint that modifies the ability of eosinophils to regulate metabolic outcomes. Generation of Il5