MilliporeSigma
  • Molecular impact of selective NFKB1 and NFKB2 signaling on DLBCL phenotype.
Molecular impact of selective NFKB1 and NFKB2 signaling on DLBCL phenotype.
Oncogene (2017-04-04)
X Guo, J L Koff, A B Moffitt, M Cinar, S Ramachandiran, Z Chen, J M Switchenko, M Mosunjac, S G Neill, K P Mann, M Bagirov, Y Du, Y Natkunam, H J Khoury, M R Rossi, W Harris, C R Flowers, I S Lossos, L H Boise, S S Dave, J Kowalski, L Bernal-Mizrachi
ABSTRACT
Diffuse large B-cell lymphoma (DLBCL) has been categorized into two molecular subtypes that have prognostic significance, namely germinal center B-cell like (GCB) and activated B-cell like (ABC). Although ABC-DLBCL has been associated with NF-κB activation, the relationships between activation of specific NF-κB signals and DLBCL phenotype remain unclear. Application of novel gene expression classifiers identified two new DLBCL categories characterized by selective p100 (NF-κB2) and p105 (NF-κB1) signaling. Interestingly, our molecular studies showed that p105 signaling is predominantly associated with GCB subtype and histone mutations. Conversely, most tumors with p100 signaling displayed ABC phenotype and harbored ABC-associated mutations in genes such as MYD88 and PIM1. In vitro, MYD88 L265P mutation promoted p100 signaling through TAK1/IKKα and GSK3/Fbxw7a pathways, suggesting a novel role for this protein as an upstream regulator of p100. p100 signaling was engaged during activation of normal B cells, suggesting p100's role in ABC phenotype development. Additionally, silencing p100 in ABC-DLBCL cells resulted in a GCB-like phenotype, with suppression of Blimp, IRF4 and XBP1 and upregulation of BCL6, whereas introduction of p52 or p100 into GC cells resulted in differentiation toward an ABC-like phenotype. Together, these findings identify specific roles for p100 and p105 signaling in defining DLBCL molecular subtypes and posit MYD88/p100 signaling as a regulator for B-cell activation.