MilliporeSigma
  • The clinical importance of tumour-infiltrating macrophages and dendritic cells in periampullary adenocarcinoma differs by morphological subtype.
The clinical importance of tumour-infiltrating macrophages and dendritic cells in periampullary adenocarcinoma differs by morphological subtype.
Journal of translational medicine (2017-07-05)
Sebastian Lundgren, Emelie Karnevi, Jacob Elebro, Björn Nodin, Mikael C I Karlsson, Jakob Eberhard, Karin Leandersson, Karin Jirström
ABSTRACT
Dendritic cells (DC) and tumour-associated macrophages (TAM) are essential in linking the innate and adaptive immune response against tumour cells and tumour progression. These cells are also potential target for immunotherapy as well as providing a handle to investigate immune status in the tumour microenvironment. The aim of the present study was to examine their impact on prognosis and chemotherapy response in periampullary adenocarcinoma, including pancreatic cancer, with particular reference to morphological subtype. The density of tolerogenic immature CD1a High density of CD1a The results from this study demonstrate that high infiltration of tolerogenic immature DCs independently predicts a shorter survival in patients with PB-type periampullary adenocarcinoma, and that high density of the MARCO