MilliporeSigma
  • Natural Prenylchalconaringenins and Prenylnaringenins as Antidiabetic Agents: α-Glucosidase and α-Amylase Inhibition and in Vivo Antihyperglycemic and Antihyperlipidemic Effects.
Natural Prenylchalconaringenins and Prenylnaringenins as Antidiabetic Agents: α-Glucosidase and α-Amylase Inhibition and in Vivo Antihyperglycemic and Antihyperlipidemic Effects.
Journal of agricultural and food chemistry (2017-01-31)
Hua Sun, Dong Wang, Xiaotong Song, Yazhou Zhang, Weina Ding, Xiaolin Peng, Xiaoting Zhang, Yashan Li, Ying Ma, Runling Wang, Peng Yu
ABSTRACT
Inhibition of α-glucosidase and α-amylase decreases postprandial blood glucose levels and delays glucose absorption, making it a treatment strategy for type 2 diabetes. This study examined in vivo and in vitro antidiabetic activities of natural prenylchalconaringenins 1 and 2 and prenylnaringenins 3 and 4, found in hops and beer. 3'-Geranylchalconaringenin (2) competitively and irreversibly inhibited α-glucosidase (IC