MilliporeSigma
PerC Manipulates Metabolism and Surface Antigens in Enteropathogenic
Frontiers in cellular and infection microbiology (2017-02-23)
Jay L Mellies, Amy Platenkamp, Jossef Osborn, Lily Ben-Avi
ABSTRACT
Enteropathogenic
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate and phenol red, powder, suitable for cell culture