MilliporeSigma
Isolation of circulating microRNA associated with RNA-binding protein.
Methods in molecular biology (Clifton, N.J.) (2013-05-31)
Andrey Turchinovich, Ludmila Weiz, Barbara Burwinkel
ABSTRACT
Circulating microRNAs (miRNAs) have been recently detected in extracellular body fluids and proved themselves as promising biomarkers for a broad spectrum of diseases. The techniques to isolate, detect, and characterize extracellular miRNAs vary significantly from report to report. In this chapter we describe a consistent, efficient and highly reproducible protocol for isolation and detection of microvesicles-free circulating miRNA from human blood plasma or cell culture media. Furthermore, since exosomes-free circulating miRNAs are associated with Argonaut proteins (the same proteins to which miRNAs are bound inside the cell), we provide a protocol for immunoprecipitation of AGO2 associated miRNAs.