MilliporeSigma
  • Vector alkaline phophatase substrate Blue III: one substrate for brightfield histochemistry and high-resolution fluorescence imaging by confocal laser scanning microscopy.
Vector alkaline phophatase substrate Blue III: one substrate for brightfield histochemistry and high-resolution fluorescence imaging by confocal laser scanning microscopy.
The Histochemical journal (1998-10-29)
M Hollinshead, J Sanderson, D J Vaux
ABSTRACT
A number of histochemical chromogenic substrates for alkaline phosphatase are commercially available and give reaction products with a range of colours for brightfield examination. Some of these reaction products are also fluorescent, exhibiting a wide excitation range and a broad emission peak. We report here that one of these substrates, Vector Blue III, yields a stable, strongly fluorescent reaction product with an excitation peak around 500 nm and a large Stokes shift to an emission peak at 680 nm. The reaction product can be excited using a mercury lamp with a fluorescein excitation filter or an argon ion laser at 488 nm or 568 nm, and the emission detected using a long-pass filter designed for Cy-5. Thus, a single substrate is suitable for brightfield imaging of tissue sections and high-resolution analysis of subcellular detail, using a confocal laser scanning microscope, in the same specimen.