MilliporeSigma
  • Synthesis and evaluation of cell-permeable biotinylated PU-H71 derivatives as tumor Hsp90 probes.
Synthesis and evaluation of cell-permeable biotinylated PU-H71 derivatives as tumor Hsp90 probes.
Beilstein journal of organic chemistry (2013-04-26)
Tony Taldone, Anna Rodina, Erica M DaGama Gomes, Matthew Riolo, Hardik J Patel, Raul Alonso-Sabadell, Danuta Zatorska, Maulik R Patel, Sarah Kishinevsky, Gabriela Chiosis
ABSTRACT
The attachment of biotin to a small molecule provides a powerful tool in biology. Here, we present a systematic approach to identify biotinylated analogues of the Hsp90 inhibitor PU-H71 that are capable of permeating cell membranes so as to enable the investigation of Hsp90 complexes in live cells. The identified derivative 2g can isolate Hsp90 through affinity purification and, as we show, represents a unique and useful tool to probe tumor Hsp90 biology in live cells by affinity capture, flow cytometry and confocal microscopy. To our knowledge, 2g is the only reported biotinylated Hsp90 probe to have such combined characteristics.