MilliporeSigma
TNFalpha induces HIF-1alpha expression through activation of IKKbeta.
Biochemical and biophysical research communications (2009-09-22)
Hsu-Ping Kuo, Dung-Fang Lee, Weiya Xia, Yongkun Wei, Mien-Chie Hung
ABSTRACT
The transcription factor hypoxia-inducible factor 1alpha (HIF-1alpha) is regulated by oxygen availability as well as various inflammatory mediators, including tumor necrosis factor alpha (TNFalpha). Early work suggested that the phosphatidylinositol-3-kinase (PI3K) and mitogen-activated protein kinase (MAPK) signaling pathways are involved in TNFalpha-mediated HIF-1alpha accumulation and activation under normoxic conditions. Here, we provide evidence showing that IkappaB kinase beta (IKKbeta) is required for HIF-1alpha regulation by TNFalpha. We found that TNFalpha enhances HIF-1alpha protein expression in various breast cancer cell lines under either normoxic or hypoxia-mimicking conditions, but has little effect on the HIF-1alpha mRNA level. Increased HIF-1alpha expression was found in IKKbeta stable clones and transient transfectants, and depletion of IKKbeta consistently reduced the amount of HIF-1alpha protein. Treatment of cells with the IKKbeta inhibitor Bay 11-7082 reduced the TNFalpha-induced HIF-1alpha expression, suggesting that IKKbeta is required in this signaling pathway. Decreased expression of vascular endothelial growth factor (VEGF), a direct target of HIF-1alpha, was shown in IKKbeta-knockout mouse embryonic fibroblast cells. We further demonstrated a positive correlation between IKKbeta and VEGF expression in primary human breast cancer specimens. Our findings indicate that TNFalpha-induced HIF-1alpha accumulation is IKKbeta dependent, and may enable further understanding of the HIF-1alpha regulation by inflammatory signals.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Roche
Anti-HA (12CA5), from mouse IgG2bκ