MilliporeSigma
  • Detection of disease-associated α-synuclein in the cerebrospinal fluid: a feasibility study.
Detection of disease-associated α-synuclein in the cerebrospinal fluid: a feasibility study.
Clinical neuropathology (2014-08-19)
Ursula Unterberger, Ingolf Lachmann, Till Voigtländer, Walter Pirker, Anna S Berghoff, Katharina Flach, Uta Wagner, Aline Geneste, Armand Perret-Liaudet, Gabor G Kovacs
ABSTRACT
With the aim to evaluate the significance and reliability of detecting disease-specific α-synuclein in the cerebrospinal fluid (CSF) we developed an ELISA and bead-assay. We used a commercial antibody (5G4) that does not bind to the physiological monomeric form of α-synuclein, but is highly specific for the disease-associated forms, including high molecular weight fraction of β-sheet rich oligomers. We applied both tests in CSF from a series of neuropathologically confirmed α-synucleinopathy cases, including Parkinson' disease dementia (PDD) and dementia with Lewy bodies (DLB) (n = 7), as well as Alzheimer' disease (n = 6), and control patients without neurodegenerative pathologies (n = 9). Disease-specific α-synuclein was detectable in the CSF in a subset of patients with α-synuclein pathology in the brain. When combined with the analysis of total α-synuclein, the bead-assay for disease-specific α-synuclein was highly specific for PDD/DLB. Detection of disease-associated αsynuclein combined with the total levels of α-synuclein is a promising tool for the in-vivo diagnosis of α-synucleinopathies, including PDD and LBD.