MilliporeSigma
  • Combination PPARgamma and RXR Agonist Treatment in Melanoma Cells: Functional Importance of S100A2.
Combination PPARgamma and RXR Agonist Treatment in Melanoma Cells: Functional Importance of S100A2.
PPAR research (2009-10-28)
Joshua P Klopper, Vibha Sharma, Reid Bissonnette, Bryan R Haugen
ABSTRACT
Nuclear hormone receptors, including RXR and PPARgamma, represent novel therapeutic targets in melanoma. We have previously shown that the DRO subline of the amelanotic melanoma A375 responds to rexinoid and thiazolidinedione (TZD) treatment in vitro and in vivo. We performed microarray analysis of A375(DRO) after TZD and combination rexinoid/TZD treatment in which the calcium binding protein S100A2 had increased expression after rexinoid or TZD treatment and a synergistic increase to combination treatment. Increased S100A2 expression is dependent on an intact PPARgamma receptor, but it is not sufficient to mediate the antiproliferative effects of rexinoid/TZD treatment. Over expression of S100A2 enhanced the effect of rexinoid and TZD treatment while inhibition of S100A2 expression attenuated the response to rexinoid/TZD treatment, suggesting that S100A2 is necessary for optimal response to RXR and PPARgamma activation by respective ligands. In summary, we have identified potential downstream mediators of rexinoid and TZD treatment in a poorly differentiated melanoma and found that alterations in S100A2 expression affect RXR and PPARgamma signaling in A375(DRO) cells. These studies provide insight into potential mechanisms of tumor response or resistance to these novel therapies.