MilliporeSigma
  • Expression of TNF-receptor associated factor 2 correlates with poor progression-free survival time in ABC-like primary nodal diffuse large B-cell lymphomas.
Expression of TNF-receptor associated factor 2 correlates with poor progression-free survival time in ABC-like primary nodal diffuse large B-cell lymphomas.
Histopathology (2008-03-04)
J C van Galen, J J F Muris, C P E Giroth, W Vos, G J Ossenkoppele, C J L M Meijer, J J Oudejans
ABSTRACT
Tumour necrosis factor (TNF)-receptor associated factor 2 (TRAF2) is an adaptor molecule involved in nuclear factor (NF)-kappaB activation, which is characteristic of in vitro activated B-cell (ABC)-like diffuse large B-cell lymphomas (DLBCL) and may result in expression of anti-apoptotic genes and poor response to chemotherapy. TRAF2 also has direct anti-apoptotic properties via interference with the apoptosis signalling cascade. The aim was to determine whether TRAF2 is preferentially expressed in ABC-like DLBCL, and whether expression correlates with clinical outcome. TRAF2 was expressed in nine of 20 tested ABC-like DLBCLs and in only one of 13 tested germinal centre B-lymphocyte (GCB)-like DLBCLs. High TRAF2 expression was correlated with high International Prognostic Index at time of presentation, high chance of relapse and short progression-free survival time in 44 tested DLBCLs. Furthermore, when analysis was restricted to ABC-like DLBCL only, TRAF2 expression was significantly associated with poor progression-free survival time. TRAF2 might be involved in activation of NF-kappaB in a subset of ABC-like DLBCL, and its expression is associated with a particularly poor outcome in primary nodal DLBCL patients. Because of its possible effect on to chemotherapy resistance, resistance, TRAF2 might be an attractive candidate as a molecular target for TRAF2+ DLBCL.