MilliporeSigma
  • Dioxin induces a novel nuclear factor, DIF-3, that is implicated in spermatogenesis.
Dioxin induces a novel nuclear factor, DIF-3, that is implicated in spermatogenesis.
FEBS letters (2001-12-01)
T Ohbayashi, K Oikawa, R Iwata, A Kameta, K Evine, T Isobe, Y Matsuda, J Mimura, Y Fujii-Kuriyama, M Kuroda, K Mukai
ABSTRACT
2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD; dioxin), a member of a class of environmental pollutants represented by polychlorinated dibenzo-p-dioxins and dibenzofurans, is one of the most toxic artificial compounds ever developed. In this study, we identified a novel TCDD target gene, DIF-3 (dioxin inducible factor-3), by cDNA representational difference analysis. DIF-3 protein is a nuclear factor and possesses a zinc-finger motif at its N-terminus. High DIF-3 mRNA expression in the testes was demonstrated by Northern blot analysis and abundant DIF-3 protein was detected during spermatogenesis. Thus, these results suggest that DIF-3 may be a target gene mediating the reproductive toxicity induced by TCDD.