MilliporeSigma
  • The use of knockout mice reveals a synergistic role of the Vav1 and Rasgrf2 gene deficiencies in lymphomagenesis and metastasis.
The use of knockout mice reveals a synergistic role of the Vav1 and Rasgrf2 gene deficiencies in lymphomagenesis and metastasis.
PloS one (2009-12-17)
Sergio Ruiz, Eugenio Santos, Xosé R Bustelo
ABSTRACT
Vav1 and RasGRF2 are GDP/GTP exchange factors for Ras superfamily GTPases with roles in the development and/or effector functions of T-lymphocytes. Given that the phenotype of Vav1(-/-), Rasgrf2(-/-) and Vav1(-/-);Rasgrf2(-/-) mice has been studied so far in young animals, we decided to explore the long-term consequences of the inactivation of those loci in the immune system. Unexpectedly, our studies revealed that the inactivation of the Vav1 proto-oncogene favors the formation of lymphoblastic lymphoma-like tumors in aging mice. Those tumors, that can be found either localized exclusively inside the thymus or widely disseminated in hematopoietic and non-hematopoietic tissues, are composed of CD3(+) lymphoblasts that display heterogeneous combinations of CD4 and CD8 surface markers. Interestingly, the additional deletion of the Rasgrf2 gene induces a shortening in the latency period for the development of those tumors, an increase in the percentage of disseminated tumors outside the thymus and, as a result, higher mortality rates. These data reveal unexpected negative roles for Vav1 and RasGRF2 in different stages of T-cell lymphoma progression. They also suggest that the inactivation of Vav1 function may represent an inadequate strategy to treat T-cell lymphomas, especially those associated with low levels of Rasgrf2 gene expression.