MilliporeSigma
  • Sham surgery and inter-individual heterogeneity are major determinants of monocyte subset kinetics in a mouse model of myocardial infarction.
Sham surgery and inter-individual heterogeneity are major determinants of monocyte subset kinetics in a mouse model of myocardial infarction.
PloS one (2014-06-04)
Jedrzej Hoffmann, Manuel Ospelt, Christian Troidl, Sandra Voss, Christoph Liebetrau, Won-Keun Kim, Andreas Rolf, Astrid Wietelmann, Thomas Braun, Kerstin Troidl, Sakthivel Sadayappan, David Barefield, Christian Hamm, Holger Nef, Helge Möllmann
ABSTRACT
Mouse models of myocardial infarction (MI) are commonly used to explore the pathophysiological role of the monocytic response in myocardial injury and to develop translational strategies. However, no study thus far has examined the potential impact of inter-individual variability and sham surgical procedures on monocyte subset kinetics after experimental MI in mice. Our goal was to investigate determinants of systemic myeloid cell subset shifts in C57BL/6 mice following MI by developing a protocol for sequential extensive flow cytometry (FCM). Following cross-sectional multiplex FCM analysis we provide for the first time a detailed description of absolute quantities, relative subset composition, and biological variability of circulating classical, intermediate, and non-classical monocyte subsets in C57BL/6 mice. By using intra-individual longitudinal measurements after MI induction, a time course of classical and non-classical monocytosis was recorded. This approach disclosed a significant reduction of monocyte subset dispersion across all investigated time points following MI. We found that in the current invasive model of chronic MI the global pattern of systemic monocyte kinetics is mainly determined by a nonspecific inflammatory response to sham surgery and not by the extent of myocardial injury. Application of sequential multiplexed FCM may help to reduce the impact of biological variability in C57BL/6 mice. Furthermore, the confounding influence of sham surgical procedures should always be considered when measuring monocyte subset kinetics in a murine model of MI.