MilliporeSigma
  • Impaired interferon-alpha production by plasmacytoid dendritic cells after cord blood transplantation in children: implication for post-transplantation toll-like receptor ligand-based immunotherapy.
Impaired interferon-alpha production by plasmacytoid dendritic cells after cord blood transplantation in children: implication for post-transplantation toll-like receptor ligand-based immunotherapy.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation (2014-08-17)
Emily Charrier, Paulo Cordeiro, Rose-Marie Brito, Michaël Harnois, Samira Mezziani, Sabine Herblot, Françoise Le Deist, Michel Duval
ABSTRACT
Plasmacytoid dendritic cells (pDCs) initiate both innate and adaptive immune responses, making them attractive targets for post-transplantation immunotherapy, particularly after cord blood transplantation (CBT). Toll-like receptor (TLR) agonists are currently studied for pDC stimulation in various clinical settings. Their efficacy depends on pDC number and functionality, which are unknown after CBT. We performed a longitudinal study of pDC reconstitution in children who underwent bone marrow transplantation (BMT) and single-unit CBT. Both CBT and unrelated BMT patients received antithymocyte globulin as part of their graft-versus-host disease prophylaxis regimen. pDC blood counts were higher in CBT patients than in healthy volunteers from 2 to 9 months after transplantation, whereas they remained lower in BMT patients. We showed that cord blood progenitors gave rise in vitro to a 500-fold increase in functional pDCs over bone marrow counterparts. Upon stimulation with a TLR agonist, pDCs from both CBT and BMT recipients upregulated T cell costimulatory molecules, whereas interferon-alpha (IFN-α) production was impaired for 9 months after CBT. TLR agonist treatment is thus not expected to induce IFN-α production by pDCs after CBT, limiting its immunotherapeutic potential. Fortunately, in vitro production of large amounts of functional pDCs from cord blood progenitors paves the way for the post-transplantation adoptive transfer of pDCs.