MilliporeSigma
  • Effects of histone deacetylase inhibitors on amygdaloid histone acetylation and neuropeptide Y expression: a role in anxiety-like and alcohol-drinking behaviours.
Effects of histone deacetylase inhibitors on amygdaloid histone acetylation and neuropeptide Y expression: a role in anxiety-like and alcohol-drinking behaviours.
The international journal of neuropsychopharmacology (2014-02-18)
Amul J Sakharkar, Huaibo Zhang, Lei Tang, Kathryn Baxstrom, Guangbin Shi, Sachin Moonat, Subhash C Pandey
ABSTRACT
Recent studies have demonstrated the involvement of epigenetic mechanisms in psychiatric disorders, including alcoholism. Here, we investigated the effects of histone deacetylase (HDAC) inhibitor, trichostatin A (TSA) on amygdaloid HDAC-induced histone deacetylation and neuropeptide Y (NPY) expression and on anxiety-like and alcohol-drinking behaviours in alcohol-preferring (P) and -non-preferring (NP) rats. It was found that P rats displayed higher anxiety-like and alcohol-drinking behaviours, higher amygdaloid nuclear, but not cytosolic, HDAC activity, which was associated with increased HDAC2 protein levels and deficits in histone acetylation and NPY expression in the central (CeA) and medial nucleus of amygdala (MeA), as compared to NP rats. TSA treatment attenuated the anxiety-like and alcohol-drinking behaviours, with concomitant reductions in amygdaloid nuclear, but not cytosolic HDAC activity, and HDAC2, but not HDAC4, protein levels in the CeA and MeA of P rats, without effect in NP rats. TSA treatment also increased global histone acetylation (H3-K9 and H4-K8) and NPY expression in the CeA and MeA of P, but not in NP rats. Histone H3 acetylation within the NPY promoter was also innately lower in the amygdala of P rats compared with NP rats; which was normalized by TSA treatment. Voluntary ethanol intake in P, but not NP rats, produced anxiolytic effects and decreased the HDAC2 levels and increased histone acetylation in the CeA and MeA. These results suggest that higher HDAC2 expression-related deficits in histone acetylation may be involved in lower NPY expression in the amygdala of P rats, and operative in controlling anxiety-like and alcohol-drinking behaviours.