MilliporeSigma
  • Neoadjuvant chemotherapy for different molecular breast cancer subtypes: a retrospective study in Russian population.
Neoadjuvant chemotherapy for different molecular breast cancer subtypes: a retrospective study in Russian population.
Medical oncology (Northwood, London, England) (2014-08-21)
Nataliya Babyshkina, Elena Malinovskaya, Stanislav Patalyak, Olga Bragina, Natalia Tarabanovskaya, Artem Doroshenko, Elena Slonimskaya, Vladimir Perelmuter, Nadejda Cherdyntseva
ABSTRACT
The aim of this retrospective study was to evaluate the objective clinical response (cOR), pathological complete response (pCR), and progression-free survival (PFS) in 231 Russian patients with four subtypes of breast cancer treated with neoadjuvant chemotherapy. About 130 (56.3 %) patients received anthracycline-based, 56 (24.2 %) capecitabine-containing (CAX), 28 (12.1 %) taxotere and 17 (7.4 %) non-anthracycline-containing chemotherapy regimens at the Tomsk Cancer Research Institute between 2000 and 2010. Tumors were subtyped according to the hormone receptor (HR) and human epidermal growth factor receptor 2 (HER2) immunohistochemical data. The majority of tumors (48.9 %) were ER+/PR+ and HER2-negative (HR+/HER2-), 10.4 % were ER+ PR+ and HER2-positive (HR+/HER2+), 9.1 % were ER-/PR- and HER2-overexpressed (HER2-enriched) and 31.6 % were ER-/PR- and HER2-negative (triple negative). Both cOR and pCR were significantly higher in the triple-negative tumors compared to the other subtypes (P = 0.021 and P = 0.033, respectively). Among the four chemotherapy regimens, only CAX regimen had a predictive value for cOR (HR 2.30, 95 % CI 1.16-4.58, P = 0.009). Multivariate regression analysis showed that the triple-negative subtype (HR 2.54, 95 % CI 1.06-1.42, P = 0.011) and CAX regimen (HR 3.01, 95 % CI 1.01-1.46, P = 0.002) were significantly associated with cOR. No association between patient's PFS and a tumor subtype was observed. However, there was a trend for a prolonged PFS among patients with cOR (P = 0.056). Our data indicate a potentially better prognosis for triple-negative breast cancer patients if treated with the CAX neoadjuvant regimen.