MilliporeSigma
  • Prognostic and treatment predictive significance of SATB1 and SATB2 expression in pancreatic and periampullary adenocarcinoma.
Prognostic and treatment predictive significance of SATB1 and SATB2 expression in pancreatic and periampullary adenocarcinoma.
Journal of translational medicine (2014-10-18)
Jacob Elebro, Margareta Heby, Alexander Gaber, Björn Nodin, Liv Jonsson, Richard Fristedt, Mathias Uhlén, Karin Jirström, Jakob Eberhard
ABSTRACT
Pancreatic cancer and other pancreaticobiliary type periampullary adenocarcinomas have a dismal prognosis even after resection and neoadjuvant chemotherapy. Intestinal type periampullary adenocarcinomas generally have a better prognosis, but little is known on optimal neoadjuvant and adjuvant treatment. New prognostic and treatment predictive biomarkers are needed for improved treatment stratification of patients with both types of periampullary adenocarcinoma. Expression of the Special AT-rich sequence-binding protein 1 (SATB1) has been demonstrated to confer a worse prognosis in several tumour types, whereas its close homologue SATB2 is a proposed diagnostic and favourable prognostic marker for colorectal cancer. The prognostic value of SATB1 and SATB2 expression in periampullary adenocarcinoma has not yet been described. Immunohistochemical expression of SATB1 and SATB2 was analysed in tissue microarrays with primary tumours and a subset of paired lymph node metastases from 175 patients operated with pancreaticoduodenectomy for periampullary adenocarcinoma. Kaplan-Meier and Cox regression analysis were applied to explore the impact of SATB1 and SATB2 expression on recurrence free survival (RFS) and overall survival (OS). Positive expression of SATB1 was denoted in 16/106 primary pancreatobiliary type tumours and 11/65 metastases, and in 15/63 primary intestinal type tumours and 4/26 metastases, respectively. Expression of SATB1 was an independent predictor of a significantly shorter RFS and OS in pancreatobiliary type, but not in intestinal type adenocarcinomas. Moreover, SATB1 expression predicted an improved response to adjuvant chemotherapy in both tumour types. SATB2-expression was seen in 3/107 pancreatobiliary type primary tumours, and in 8/61 intestinal type primary tumours. The small number of cases with positive SATB2 expression did not allow for any firm conclusions on its prognostic value. These findings demonstrate the potential utility of SATB1 as a prognostic and predictive biomarker for chemotherapy response in both intestinal type and pancreatobiliary type periampullary adenocarcinomas, including pancreatic cancer.