MilliporeSigma
  • Risk of antibody-mediated rejection in kidney transplant recipients with anti-HLA-C donor-specific antibodies.
Risk of antibody-mediated rejection in kidney transplant recipients with anti-HLA-C donor-specific antibodies.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2014-05-09)
O Aubert, M-C Bories, C Suberbielle, R Snanoudj, D Anglicheau, M Rabant, F Martinez, A Scemla, C Legendre, R Sberro-Soussan
ABSTRACT
Anti-HLA donor-specific antibodies (DSAs) cause acute and chronic antibody-mediated rejection (AMR). However, the clinical relevance of anti-HLA-C antibodies remains unclear. We evaluated the clinical relevance of the presence of anti-HLA-C DSA at day 0 in renal transplant recipients. In this retrospective, case-controlled study, 608 patients who underwent kidney transplantation between August 2008 and March 2012 were screened for the presence of isolated anti-HLA-C DSA at day 0. A total of 22 renal transplant recipients were selected and followed for a period of 1 year. AMR was classified according to the Banff classification. The 22 patients were compared with 88 immunized patients. Acute AMR was diagnosed in six patients (27.3%). The median level of DSA at day 0 was 1179 (530-17,941). The mean fluorescence intensity in the anti-C group was 4966 (978-17,941) in the AMR group and 981 (530-8012) in the group of patients without AMR. Acute AMR was diagnosed less frequently in the 88 immunized individuals (9.1%) than in the DSA anti-C group (p = 0.033). The level of DSA at day 0 was predictive for AMR (p = 0.017). Patients with a high level of pretransplant anti-HLA-C DSAs are likely to develop acute AMR during the first year after transplantation.