MilliporeSigma
Regulation of the VHL/HIF-1 pathway by DJ-1.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-06-06)
Mohammad Parsanejad, Yi Zhang, Dianbo Qu, Isabella Irrcher, Maxime W C Rousseaux, Hossein Aleyasin, Fatemeh Kamkar, Steve Callaghan, Ruth S Slack, Tak W Mak, Stephen Lee, Daniel Figeys, David S Park
ABSTRACT
DJ-1 (PARK7) is a gene linked to autosomal recessive Parkinson disease (PD). We showed previously that DJ-1 loss sensitizes neurons in models of PD and stroke. However, the biochemical mechanisms underlying this protective role are not completely clear. Here, we identify Von Hippel Lindau (VHL) protein as a critical DJ-1-interacting protein. We provide evidence that DJ-1 negatively regulates VHL ubiquitination activity of the α-subunit of hypoxia-inducible factor-1 (HIF-1α) by inhibiting HIF-VHL interaction. Consistent with this observation, DJ-1 deficiency leads to lowered HIF-1α levels in models of both hypoxia and oxidative stress, two stresses known to stabilize HIF-1α. We also demonstrate that HIF-1α accumulation rescues DJ-1-deficient neurons against 1-methyl-4-phenylpyridinium-induced toxicity. Interestingly, lymphoblast cells extracted from DJ-1-related PD patients show impaired HIF-1α stabilization when compared with normal individuals, indicating that the DJ-1-VHL link may also be relevant to a human context. Together, our findings delineate a model by which DJ-1 mediates neuronal survival by regulation of the VHL-HIF-1α pathway.