MilliporeSigma
  • Involvement of amygdaloid neuropeptide Y in the anxiolytic effects of acupuncture during ethanol withdrawal in rats.
Involvement of amygdaloid neuropeptide Y in the anxiolytic effects of acupuncture during ethanol withdrawal in rats.
Neuroscience letters (2014-03-29)
Zhenglin Zhao, Sang Chan Kim, Yiyan Wu, Jie Zhang, Yanji Xu, Il Je Cho, Chae Ha Yang, Bong Hyo Lee, Rongjie Zhao
ABSTRACT
The role of neuropeptide Y (NPY) in the central nucleus of amygdala (CeA) in the preventive effects of acupuncture against ethanol withdrawal-induced anxiety was investigated. Rats were treated with 3g/kg/day of ethanol for 28 days, followed by 3 days of withdrawal. Bilateral acupuncture treatment at HT7 (Shen-Men), PC6 (Nei-Guan) or a non-acupoint was respectively added to the rats during the withdrawal once a day for three days. Enzyme-linked immunosorbent assays and real-time polymerase chain reaction analyses showed there was a significant decrease in NPY protein and mRNA expression in the CeA during ethanol withdrawal, which was reversed by acupuncture at HT7 but neither at PC6 nor at a non-acupoint. Acupuncture at HT7 also greatly inhibited the decrease in cAMP response element-binding protein (CREB) phosphorylation in the CeA. In elevated plus maze tests, a selective NPY Y1 receptor antagonist BIBP 3226 into the CeA before the acupuncture abolished almost completely the anxiolytic effect of acupuncture at HT7. These results suggest that acupuncture at HT7 rescues the depletion of amygdaloid NPY and reverses the decrease in CREB phosphorylation to produce anxiolytic effects during ethanol withdrawal.