MilliporeSigma
  • 1,3-Dipolar cycloaddition of arynes with azomethine imines: synthesis of 1,2-dihydropyrazolo[1,2-a]indazol-3(9H)-ones.
1,3-Dipolar cycloaddition of arynes with azomethine imines: synthesis of 1,2-dihydropyrazolo[1,2-a]indazol-3(9H)-ones.
Tetrahedron letters (2010-02-18)
Feng Shi, Raffaella Mancuso, Richard C Larock
ABSTRACT
A [3+2] 1,3-dipolar cycloaddition reaction of arynes with stable azomethine imines has been developed. The reaction rapidly assembles tricyclic pyrazoloindazolone derivatives in moderate yields under mild reaction conditions.