MilliporeSigma
  • Overexpression of metastasis-associated protein 2 is associated with hepatocellular carcinoma size and differentiation.
Overexpression of metastasis-associated protein 2 is associated with hepatocellular carcinoma size and differentiation.
Journal of gastroenterology and hepatology (2009-08-26)
Hyunseung Lee, Soo Hyung Ryu, Soon Sun Hong, Dong Dae Seo, Hyun Joo Min, Myoung Kuk Jang, Ho Jeong Kwon, Ensil Yu, Young-Hwa Chung, Kyu-Won Kim
ABSTRACT
Metastasis is a multistep event in which neoplastic cells detach from the tumor, migrate, disseminate, extravasate, and eventually proliferate at the secondary distant sites. Hepatocellular carcinoma (HCC) is characterized by hypervascularity and frequent metastasis. Recently, metastasis-associated proteins were identified and named metastatic tumor antigens (MTA) 1, 2, and 3. They have been found to be contained in the nucleosome remodeling and histone deacetylase complex. MTA2 has been reported to interact with p53 and inhibit p53-mediated cell growth arrest and apoptosis by deacetylation. Although it has been reported that the expression of MTA1 is related to tumor progression and metastasis, it is still unclear how MTA2 is involved in HCC. In this study, we found that the overexpression of MTA2 is associated with HCC size and differentiation after hepatectomy. The expression of MTA2 was examined in 506 human HCC samples that underwent hepatic resection using tissue microarray. The expression of MTA2 was classified into 0, 1, 2, and 3, based on immunoreactivity. The expression of MTA2 was predominantly localized to the nucleus. MTA2 was detected in 487 (96.2%) of the 506 human HCC samples. Notably, the MTA2 expression level strongly increased depending on the size and differentiation of HCC. These findings indicate a tight correlation between the MTA2 expression level and HCC size and differentiation. Therefore, MTA2 might be a predictor of aggressive phenotypes and a possible target molecule for anticancer drug design in human HCC.