MilliporeSigma
  • The overexpression of KIT proto-oncogene in acute leukemic cells is not necessarily caused by the gene mutation.
The overexpression of KIT proto-oncogene in acute leukemic cells is not necessarily caused by the gene mutation.
Acta haematologica (2014-09-24)
Damian Szatkowski, Andrzej Hellmann
ABSTRACT
KIT is detected in a variety of cells, also in acute leukemia. Inhibition of wild-type KIT is not always satisfactory. The aim of this work was to evaluate the frequency of the most common KIT mutations in acute myeloid leukemia (AML) and determine the correlation between mutation and expression level. Samples were obtained from 75 patients with AL. CD117 presence was shown in 45 of 51 patients with AML and in 1 of 16 patients with acute lymphocytic leukemia (ALL). Asp816Val mutation was found in 3.5% of cases of AML and Val560Gly mutation in 1 sample with acute biclonal leukemia. Other genetic changes were found in 15 of 57 samples with AML: polymorphisms Met541Leu in 14% of cases, Lys546Lys in 7% and 1 case of acute biclonal leukemia, Ile798Ile in 5.3% of cases, Met541Leu in 1 acute biphenotypic leukemia and in 6.3% of ALL. Polymorphism Lys546Lys was also shown in 1 case of acute biclonal leukemia. Nonsilent genetic changes were detected in a total of 23% cases with core binding factor leukemia. There was no statistical significance between KIT expression and genetic changes. There was no correlation between the incidence and types of KIT mutations and its expression on cells in AML.