MilliporeSigma
  • Malignant epithelioid mesothelioma: anti-mesothelial marker expression correlates with histological pattern.
Malignant epithelioid mesothelioma: anti-mesothelial marker expression correlates with histological pattern.
Histopathology (2002-03-21)
R L Attanoos, R Webb, S D Dojcinov, A R Gibbs
ABSTRACT
Malignant epithelioid mesothelioma shows marked cytoarchitectural diversity. The aim of the study was to evaluate how immunoreactivity with mesothelial markers related to histological pattern. Ninety-two cases of malignant epithelioid mesothelioma (60 pleural, 32 peritoneal) were examined and classified as exhibiting tubulopapillary, adenomatoid, solid, small cell or pleomorphic patterns. All cases were immunohistochemically stained with thrombomodulin, calretinin, CD44H, and cytokeratin 5/6. Cases of malignant mesothelioma exhibited a number of different histological forms. Immunohistochemical expression of each mesothelial marker tested was not homogeneous across different histological patterns of malignant epithelioid mesothelioma, even within the same tumour section. Calretinin (with nuclear expression) was identified to show the highest overall sensitivity and lowest range variation in staining (67% sensitivity in small cell areas to 100% expression in pleomorphic areas). Cytokeratin 5/6 and thrombomodulin yielded similar overall sensitivity. Thrombomodulin appeared to demonstrate higher sensitivity for small cell variant tumour (83% sensitivity). A notable advantage with cytokeratin 5/6 was that expression was more diffuse in nature rather than the focal membranous elaboration seen in thrombomodulin. The widest range of staining was seen in small cell mesothelioma (83% sensitivity with thrombomodulin to 17% sensitivity with cytokeratin 5/6) and in tubulopapillary areas (90% sensitivity with calretinin to 38% sensitivity with CD44H). Calretinin appears most useful and shows the highest overall sensitivity for malignant epithelioid mesothelioma, with good expression in areas displaying a tubulopapillary, adenomatoid, solid and pleomorphic pattern. For small cell mesothelioma, thrombomodulin appears to confer higher sensitivity and is advocated, in this setting, as the first line mesothelial marker. Cytokeratin 5/6 is a useful and easily interpretable mesothelial marker. CD44H is not of particular use in the diagnosis of malignant epithelioid mesothelioma. Accurate interpretation of immunohistochemistry in mesothelioma requires an awareness of the immunophenotypic heterogeneity identified in different histological forms of the tumour, and this is of particular importance in small biopsies.
Sigma-Aldrich
Thrombomodulin (1009) Mouse Monoclonal Antibody