MilliporeSigma
Phosphorylase b kinase inheritance in mice.
Science (New York, N.Y.) (1967-03-24)
J B Lyon, J Porter, M Robertson
ABSTRACT
The gene for phosphoryl ase b kinase, a skeletal muscle enzyme, has been located on the X chromo some of mice. The inheritance of the enzyme through two generations from original matings between one inbred strain of mice, the I, which lacks the enzyme, and another strain, the C(57,) follows the classical Mendelian pattern.