MilliporeSigma
  • Use of SE-30 as a stationary phase for the gas-liquid chromatography of drugs.
Use of SE-30 as a stationary phase for the gas-liquid chromatography of drugs.
Journal of chromatography (1975-03-12)
A C Moffat
ABSTRACT
The dimethyl silicone elastomer SE-30 has been chosen as the preferred liquid phase for the gas-liquid chromatographic analysis of drugs, and retention index data have been compiled for 480 drugs and commonly occurring chemicals such as plasticisers. The inter-laboratory variation in measurement of retention indices has been measured for three drugs in eleven laboratories and the standard deviations were between 20 and 15 retention index units.